Assay Details
Target Gene Details
Entrez Gene ID: | 17075 |
Gene Name: | epithelial cell adhesion molecule |
Gene Aliases: |
CD326, EGP, EGP-2, Egp314, Ep-CAM, EpCAM1, GA733-2, Ly74, TROP1, Tacsd1, Tacstd1, gp40 |
Location: |
Chr.17:87635979-87651127 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epcam | NM_008532.2 | 4 | 677 | NP_032558.2 |
| AK133510.1 | 3 | 359 | ||
| AK145752.1 | 3 | 253 | BAE26627.1 | |
| AK167182.1 | 3 | 322 | BAE39317.1 | |
| BC005618.1 | 3 | 318 | AAH05618.1 | |
| BC094465.1 | 3 | 343 | AAH94465.1 | |
| M76124.1 | 3 | 329 | AAA37543.1 |

Genomic Map