Assay Details
Target Gene Details
Entrez Gene ID: | 14158 |
Gene Name: | fer (fms/fps related) protein kinase |
Gene Aliases: |
AV082135, C330004K01Rik, Fert, Fert2 |
Location: |
Chr.17:63829950-64146519 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fer | NM_001037997.3 | NP_001033086.2 | ||
| AK021175.1 | BAB32315.1 | |||
| AK157828.1 | BAE34213.1 | |||
| AK165973.1 | BAE38493.1 |
More Information
Set Membership: |
|

Genomic Map