Assay Details
Target Gene Details
Entrez Gene ID: | 76438 |
Gene Name: | raftlin lipid raft linker 1 |
Gene Aliases: |
2310015N21Rik, AI853526 |
Location: |
Chr.17:49993258-50190497 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rftn1 | NM_181397.2 | NP_852062.1 | ||
| AK088466.1 | ||||
| AK152130.1 | BAE30971.1 | |||
| AK153288.1 | BAE31871.1 | |||
| AK155870.1 | BAE33473.1 | |||
| AK170913.1 | ||||
| AK172884.1 | BAD32162.1 | |||
| BC052074.1 | AAH52074.1 |

Genomic Map