Assay Details
Target Gene Details
Entrez Gene ID: | 319991 |
Gene Name: | kinesin family member 6 |
Gene Aliases: |
D130004B10Rik, D130084M03Rik |
Location: |
Chr.17:49615104-49909847 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif6 | NM_177052.3 | NP_796026.2 | ||
| AK084082.1 |
More Information
Set Membership: |
|

Genomic Map