Assay Details
Target Gene Details
Entrez Gene ID: | 74142 |
Gene Name: | lon peptidase 1, mitochondrial |
Gene Aliases: |
1200017E13Rik, LON, Prss15 |
Location: |
Chr.17:56614297-56626903 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lonp1 | NM_028782.2 | 10 | 1597 | NP_083058.2 |
| AK004820.1 | 10 | 1581 | BAB23591.1 | |
| AK131910.1 | 10 | 1583 | BAE20868.1 | |
| AK157474.1 | 10 | 1576 | BAE34094.1 | |
| AK159302.1 | 10 | 1582 | BAE34972.1 | |
| AK160071.1 | 10 | 1582 | BAE35606.1 | |
| AK160076.1 | 10 | 1583 | ||
| AK161983.1 | 10 | 1582 | BAE36666.1 | |
| AK189995.1 | ||||
| AK193246.1 | 2 | 187 | ||
| AK193735.1 | ||||
| AK203362.1 | 3 | 283 | ||
| AY190302.1 | 10 | 1567 | AAN85210.1 | |
| BC092212.1 | AAH92212.1 |
More Information
Set Membership: |
|

Genomic Map