Assay Details
Target Gene Details
Entrez Gene ID: | 106794 |
Gene Name: | DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57 |
Gene Aliases: |
AW494914 |
Location: |
Chr.17:80238304-80290476 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhx57 | NM_001163759.1 | 1 | 290 | NP_001157231.1 |
| NM_198942.2 | 1 | 290 | NP_945180.2 | |
| BC062952.1 | 1 | 290 | AAH62952.1 |
Target Gene Details
Entrez Gene ID: | 378462 |
Gene Name: | MORN repeat containing 2 |
Gene Aliases: |
Mopt |
Location: |
Chr.17:80290212-80305691 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Morn2 | NM_194269.2 | NP_919245.1 | ||
| AY367765.1 | AAQ73169.1 |
More Information
Set Membership: |
|

Genomic Map