Assay Details
Target Gene Details
Entrez Gene ID: | 18207 |
Gene Name: | nth (endonuclease III)-like 1 (E.coli) |
Gene Aliases: |
Nth1, Octs3 |
Location: |
Chr.17:24632682-24638838 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nthl1 | NM_008743.2 | NP_032769.2 | ||
| AB006812.1 | BAA22080.1 | |||
| AK033701.1 | ||||
| AV377313.2 | ||||
| BC138852.1 | ||||
| BC138855.1 | ||||
| BC145443.1 | ||||
| CJ211561.1 | ||||
| CX565655.1 | ||||
| Y09688.1 | CAA70866.1 |
More Information
Set Membership: |
|

Genomic Map