Assay Details
Target Gene Details
Entrez Gene ID: | 51792 |
Gene Name: | protein phosphatase 2, regulatory subunit A, alpha |
Gene Aliases: |
6330556D22Rik, PP2A, PR65 |
Location: |
Chr.17:20945454-20965905 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2r1a | NM_016891.3 | NP_058587.1 | ||
| AB021743.1 | BAA75478.1 | |||
| AK054239.1 | BAC35700.1 | |||
| AK078135.1 | BAC37143.1 | |||
| AK088776.1 | BAC40565.1 | |||
| AK178876.1 | ||||
| AK206743.1 | ||||
| BC006606.1 | AAH06606.1 | |||
| BC052678.1 | AAH52678.1 |
More Information
Set Membership: |
|

Genomic Map