Assay Details
Target Gene Details
Entrez Gene ID: | 15013 |
Gene Name: | histocompatibility 2, Q region locus 2 |
Gene Aliases: |
BE136769, ENSMUSG00000079507, Gm11132, H-2Q2, gs14-2 |
Location: |
Chr.17:35342120-35346764 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-Q2 | NM_010392.2 | NP_034522.2 | ||
| AY989880.1 | AAY85367.1 | |||
| BC138688.1 | ||||
| BC138689.1 |
More Information
Set Membership: |
|

Genomic Map