Assay Details
Target Gene Details
Entrez Gene ID: | 74142 |
Gene Name: | lon peptidase 1, mitochondrial |
Gene Aliases: |
1200017E13Rik, LON, Prss15 |
Location: |
Chr.17:56614297-56626903 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lonp1 | NM_028782.2 | NP_083058.2 | ||
| AK004820.1 | BAB23591.1 | |||
| AK131910.1 | BAE20868.1 | |||
| AK157474.1 | BAE34094.1 | |||
| AK159302.1 | BAE34972.1 | |||
| AK160071.1 | BAE35606.1 | |||
| AK160076.1 | ||||
| AK161983.1 | BAE36666.1 | |||
| AK187055.1 | ||||
| AY190302.1 | AAN85210.1 |
More Information
Set Membership: |
|

Genomic Map