Assay Details
Target Gene Details
Entrez Gene ID: | 15043 |
Gene Name: | histocompatibility 2, T region locus 3 |
Gene Aliases: |
H-2T3, H2-Tw3, TL |
Location: |
Chr.17:36185571-36190151 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H2-T3 | NM_008208.4 | NP_032234.3 | ||
| AK033602.1 | BAC28383.1 | |||
| AY989874.1 | AAY85361.1 | |||
| AY989875.1 | AAY85362.1 | |||
| AY989876.1 | AAY85363.1 | |||
| AY989877.1 | AAY85364.1 | |||
| AY989878.1 | AAY85365.1 | |||
| BC125378.1 | ||||
| BC125380.1 | ||||
| D86082.1 | BAA13015.1 |
More Information
Set Membership: |
|

Genomic Map