Assay Details
Target Gene Details
Entrez Gene ID: | 353344 |
Gene Name: | opsin 5 |
Gene Aliases: |
Gpr136, Neuropsin, PGR12, TMEM13 |
Location: |
Chr.17:42556779-42620470 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Opn5 | NM_181753.4 | NP_861418.2 | ||
| AK039525.1 | BAC30373.1 | |||
| AY318865.1 | AAR08201.1 | |||
| BC125611.1 | ||||
| BC132010.1 |
More Information
Set Membership: |
|

Genomic Map