Assay Details
Target Gene Details
Entrez Gene ID: | 240121 |
Gene Name: | fibronectin type 3 and SPRY domain-containing protein |
Gene Aliases: |
- |
Location: |
Chr.17:55986462-55996883 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fsd1 | NM_183178.2 | NP_899001.1 | ||
| AK140132.1 | BAE24249.1 | |||
| AK144257.1 | BAE25801.1 | |||
| BC055106.1 | AAH55106.1 |
More Information
Set Membership: |
|

Genomic Map