Assay Details
Target Gene Details
Entrez Gene ID: | 74764 |
Gene Name: | kinesin light chain 4 |
Gene Aliases: |
1200014P03Rik, AA408085, AW146303, Knsl8 |
Location: |
Chr.17:46630624-46646167 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klc4 | NM_029091.2 | NP_083367.1 | ||
| AK004774.1 | BAB23552.1 | |||
| AK170793.1 | BAE42032.1 | |||
| BC005746.1 | AAH05746.1 |
More Information
Set Membership: |
|

Genomic Map