Assay Details
Target Gene Details
Entrez Gene ID: | 50782 |
Gene Name: | regulator of G-protein signaling 11 |
Gene Aliases: |
C78048 |
Location: |
Chr.17:26202962-26211324 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgs11 | NM_001081069.1 | NP_001074538.1 | ||
| AF061934.1 | AAC70012.1 | |||
| AK039314.1 | ||||
| AK080707.1 | ||||
| AK161551.1 |
More Information
Set Membership: |
|

Genomic Map