Assay Details
Target Gene Details
Entrez Gene ID: | 79044 |
Gene Name: | mitochondrial ribosomal protein S34 |
Gene Aliases: |
0610007F04Rik, 5330430D13Rik, AV001970, Tce2 |
Location: |
Chr.17:24895120-24896274 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps34 | NM_023260.1 | NP_075749.1 | ||
| AK002285.1 | BAB21988.1 | |||
| AK004049.1 | BAB23142.1 | |||
| AK007337.1 | BAB24970.1 | |||
| AK019922.1 | 1 | 526 | ||
| AK216319.1 | ||||
| BC024336.1 | AAH24336.1 |
More Information
Set Membership: |
|

Genomic Map