Assay Details
Target Gene Details
Entrez Gene ID: | 18771 |
Gene Name: | Pbx/knotted 1 homeobox |
Gene Aliases: |
D17Wsu76e, PREP1 |
Location: |
Chr.17:31564721-31607693 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pknox1 | NM_016670.3 | 8 | 961 | NP_057879.2 |
| NR_027493.1 | 9 | 1208 | ||
| AF061270.1 | 8 | 933 | AAC15990.1 | |
| AK131788.1 | 7 | 990 | BAE20805.1 | |
| AK134000.1 | 9 | 1118 | BAE21975.1 | |
| BC052701.1 | 7 | 856 | AAH52701.1 |
More Information
Set Membership: |
|

Genomic Map