Assay Details
Target Gene Details
Entrez Gene ID: | 73504 |
Gene Name: | RIKEN cDNA 1700071M16 gene |
Gene Aliases: |
- |
Location: |
Chr.17:43588340-43591509 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700071M16Rik | NR_045444.1 | 3 | 2212 | |
| NR_045445.1 | 4 | 668 | ||
| AK133239.1 | 3 | 2212 | ||
| AK161361.1 | 4 | 669 | ||
| AV274656.2 | 2 | 368 |
Target Gene Details
Entrez Gene ID: | 27226 |
Gene Name: | phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma) |
Gene Aliases: |
R75400 |
Location: |
Chr.17:43568082-43612202 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g7 | NM_013737.5 | 2 | 405 | NP_038765.2 |
| AK005158.1 | 2 | 142 | BAB23849.1 | |
| AK005210.1 | 2 | 199 | ||
| AK051454.1 | 2 | 406 | BAC34647.1 | |
| AK155678.1 | 3 | 256 | BAE33386.1 | |
| BC010726.1 | 2 | 161 | AAH10726.1 | |
| U34277.1 | 2 | 151 | AAC52274.1 |
More Information
Set Membership: |
|

Genomic Map