Assay Details
Target Gene Details
Entrez Gene ID: | 193813 |
Gene Name: | multiple coagulation factor deficiency 2 |
Gene Aliases: |
1810021C21Rik, F5f8d, Lman1ip, Sdnsf |
Location: |
Chr.17:87254443-87265947 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcfd2 | NM_139295.3 | 2 | 242 | NP_647456.1 |
| NM_176808.5 | 3 | 467 | NP_789778.1 | |
| AF475283.1 | 2 | 210 | AAM28464.1 | |
| AK033139.1 | 2 | 176 | BAC28169.1 | |
| AK075781.1 | 3 | 400 | BAC35955.1 | |
| AK077139.1 | 2 | 193 | BAC36638.1 | |
| AK077631.1 | 2 | 176 | BAC36914.1 | |
| AK141599.1 | 3 | 425 | BAE24757.1 | |
| AK151803.1 | 2 | 231 | BAE30702.1 | |
| AK206943.1 | 1 | 121 | ||
| BC003996.1 | 2 | 169 | AAH03996.1 | |
| GQ896355.1 | 1 | 113 | ||
| GQ896356.1 | 1 | 113 |
More Information
Set Membership: |
|

Genomic Map