Assay Details
Target Gene Details
Entrez Gene ID: | 68342 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10 |
Gene Aliases: |
0610011B04Rik, 22kDa, PDSW |
Location: |
Chr.17:24722067-24724388 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufb10 | NM_026684.2 | 1 | 56 | NP_080960.1 |
| AK002517.1 | BAB22156.1 | |||
| AK151404.1 | 1 | 90 | BAE30371.1 | |
| BC031664.1 | 1 | 56 | AAH31664.1 | |
| BC043013.1 | AAH43013.1 |
More Information
Set Membership: |
|

Genomic Map