Assay Details
Target Gene Details
Entrez Gene ID: | 213389 |
Gene Name: | PR domain containing 9 |
Gene Aliases: |
BC012016, Dsbc1, G1-419-29, Meisetz, Rcr1, repro7 |
Location: |
Chr.17:15543072-15564379 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prdm9 | NM_144809.2 | 9 | 1123 | NP_659058.2 |
| AB844106.1 | 9 | 1123 | ||
| AB844107.1 | 9 | 1123 | ||
| AB844108.1 | 9 | 1123 | ||
| AB844109.1 | 9 | 1129 | ||
| AB844110.1 | 9 | 1123 | ||
| AB844111.1 | 9 | 1101 | ||
| AB844112.1 | 9 | 1101 | ||
| AB844113.1 | 9 | 1123 | ||
| AB844114.1 | 9 | 1123 | ||
| BC012016.1 | 11 | 1264 | AAH12016.1 | |
| BC023014.1 | 4 | 785 | ||
| BC049903.1 | 11 | 1264 | ||
| FJ212287.1 | 9 | 1101 | ||
| GU216230.1 | 9 | 1101 | ||
| HQ704390.1 | 9 | 1101 | ||
| HQ704391.1 | 9 | 1101 |
More Information
Set Membership: |
|

Genomic Map