Assay Details
Target Gene Details
Entrez Gene ID: | 17256 |
Gene Name: | male enhanced antigen 1 |
Gene Aliases: |
Mea-1 |
Location: |
Chr.17:46679138-46683126 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mea1 | NM_001277309.1 | NP_001264238.1 | ||
| NM_001277310.1 | NP_001264239.1 | |||
| NM_001277311.1 | NP_001264240.1 | |||
| NM_001277312.1 | NP_001264241.1 | |||
| NM_010787.2 | NP_034917.1 | |||
| NR_102392.1 | ||||
| AK005106.1 | BAB23822.1 | |||
| AK005369.1 | BAB23978.1 | |||
| AK008194.1 | BAB25525.1 | |||
| AK008305.1 | BAB25590.1 | |||
| AK010863.1 | BAB27231.1 | |||
| AK010904.1 | BAB27257.1 | |||
| AK010962.1 | BAB27294.1 | |||
| AK011189.1 | BAB27454.1 | |||
| AK012617.1 | BAB28358.1 | |||
| BC013344.1 | AAH13344.1 | |||
| BG085669.2 | ||||
| BY707372.1 | ||||
| D17341.1 | BAA04159.1 | |||
| L10401.1 | AAA39520.1 | |||
| M27938.1 | AAA39519.1 |

Genomic Map