Assay Details
Target Gene Details
Entrez Gene ID: | 75292 |
Gene Name: | protein kinase D3 |
Gene Aliases: |
4930557O20Rik, 5730497N19Rik, PKD3, Pkcnu, Prkcn |
Location: |
Chr.17:78949405-79020816 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkd3 | NM_001171004.1 | NP_001164475.1 | ||
| NM_001171005.1 | NP_001164476.1 | |||
| NM_029239.3 | NP_083515.2 | |||
| AK030058.1 | ||||
| BC037012.1 | AAH37012.1 | |||
| BC089164.1 | AAH89164.1 |
More Information
Set Membership: |
|

Genomic Map