Assay Details
Target Gene Details
Entrez Gene ID: | 268977 |
Gene Name: | latent transforming growth factor beta binding protein 1 |
Gene Aliases: |
9430031G15Rik, 9830146M04, Ltbp-1, Tgfb, b2b1000Clo |
Location: |
Chr.17:75005529-75392967 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 18 - Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ltbp1 | NM_019919.3 | NP_064303.2 | ||
| NM_206958.2 | NP_996841.1 | |||
| AF022889.1 | AAC33307.1 | |||
| AK036047.1 | ||||
| AK054512.1 | 3 | 424 | ||
| BC094612.1 | AAH94612.1 |
More Information
Set Membership: |
|

Genomic Map