Assay Details
Target Gene Details
Entrez Gene ID: | 19170 |
Gene Name: | proteasome (prosome, macropain) subunit, beta type 1 |
Gene Aliases: |
AA409053, C81484, Lmpc5 |
Location: |
Chr.17:15475721-15498276 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmb1 | NM_011185.3 | NP_035315.1 | ||
| AK028265.1 | ||||
| AK077520.1 | BAC36841.1 | |||
| AK132161.1 | BAE21005.1 | |||
| AK167647.1 | BAE39698.1 | |||
| BC018351.1 | AAH18351.1 | |||
| U60824.1 | AAB37251.1 | |||
| X80686.1 | CAA56701.1 |
More Information
Set Membership: |
|

Genomic Map