Assay Details
Target Gene Details
Entrez Gene ID: | 16913 |
Gene Name: | proteasome (prosome, macropain) subunit, beta type 8 (large multifunctional peptidase 7) |
Gene Aliases: |
Lmp-7, Lmp7 |
Location: |
Chr.17:34198195-34201454 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmb8 | NM_010724.2 | NP_034854.2 | ||
| AK031801.1 | BAC27555.1 | |||
| AK050627.1 | BAE20682.1 | |||
| AK146182.1 | BAE26960.1 | |||
| AK150674.1 | BAE29756.1 | |||
| AK159971.1 | BAE35524.1 | |||
| AK169753.1 | ||||
| BC013785.1 | AAH13785.1 | |||
| BC051450.1 | AAH51450.1 | |||
| KJ650228.1 | ||||
| U22031.1 | AAA75033.1 | |||
| U22032.1 | AAA75034.1 | |||
| U22033.1 | AAA75035.1 | |||
| U22034.1 | AAA75036.1 | |||
| U22035.1 | AAA75037.1 | |||
| X64449.1 | CAA45779.1 |
More Information
Set Membership: |
|

Genomic Map