Assay Details
Target Gene Details
Entrez Gene ID: | 240028 |
Gene Name: | leucyl/cystinyl aminopeptidase |
Gene Aliases: |
2010309L07Rik, 4732490P18Rik, CAP, IRAP, PLAP, gp160, vp165 |
Location: |
Chr.17:17527723-17624489 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lnpep | NM_172827.3 | NP_766415.1 | ||
| AK029094.1 | BAC26293.1 | |||
| AK036967.1 | BAC29651.1 | |||
| BC066791.1 | AAH66791.1 | |||
| BC120925.1 | ||||
| BC120926.1 |
More Information
Set Membership: |
|

Genomic Map