Assay Details
Target Gene Details
Entrez Gene ID: | 16912 |
Gene Name: | proteasome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2) |
Gene Aliases: |
Lmp-2, Lmp2 |
Location: |
Chr.17:34182030-34187330 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Psmb9 | NM_013585.2 | NP_038613.1 | ||
| AK008429.1 | BAB25664.1 | |||
| AK152743.1 | BAE31463.1 | |||
| AK171670.1 | BAE42601.1 | |||
| BC116364.2 | ||||
| CT010227.1 | CAJ18435.1 | |||
| D44454.1 | BAA22575.1 | |||
| D44456.1 | BAA22577.1 | |||
| D44457.1 | BAA22578.1 | |||
| D44458.1 | BAA22579.1 | |||
| D44461.1 | BAA22582.1 | |||
| KJ650226.1 | ||||
| S58203.1 | AAP13903.1 | |||
| S59862.1 | AAB20105.1 | |||
| U22447.1 | AAA75304.1 | |||
| U22448.1 | AAA75305.1 | |||
| U22919.1 | AAA75306.1 | |||
| U22920.1 | AAA75307.1 |
More Information
Set Membership: |
|

Genomic Map