Assay Details
Target Gene Details
Entrez Gene ID: | 66229 |
Gene Name: | ribosomal protein L7-like 1 |
Gene Aliases: |
1500016H10Rik |
Location: |
Chr.17:46773907-46782656 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl7l1 | NM_025433.3 | NP_079709.2 | ||
| AK005264.1 | ||||
| AK007884.1 | BAB25329.1 | |||
| AK008672.1 | ||||
| AK090038.1 | ||||
| AK090082.1 | BAC41084.1 | |||
| AK166499.1 | BAE38811.1 | |||
| AK167036.1 | ||||
| BC030165.1 | AAH30165.1 |
More Information
Set Membership: |
|

Genomic Map