Assay Details
Target Gene Details
Entrez Gene ID: | 71461 |
Gene Name: | PTK7 protein tyrosine kinase 7 |
Gene Aliases: |
8430404F20Rik, chz, mPTK7/CCK4 |
Location: |
Chr.17:46564451-46630049 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptk7 | NM_175168.4 | 9 | 1583 | NP_780377.1 |
| AF531873.1 | 9 | 1433 | AAQ10071.1 | |
| AK053044.1 | 9 | 1516 | BAC35248.1 | |
| AK163135.1 | 9 | 1516 | BAE37208.1 | |
| BC076578.1 | 9 | 1583 | AAH76578.1 |
More Information
Set Membership: |
|

Genomic Map