Assay Details
Target Gene Details
Entrez Gene ID: | 53901 |
Gene Name: | regulator of calcineurin 2 |
Gene Aliases: |
Csp2, Dscr1l1, MCIP2, ZAKI-4 |
Location: |
Chr.17:43801281-44039516 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcan2 | NM_207649.1 | 1 | 43 | NP_997532.1 |
| AK138768.1 | BAE23773.1 | |||
| BB635249.1 | ||||
| BY740155.1 | 1 | 50 |
More Information
Set Membership: |
|

Genomic Map