Assay Details
Target Gene Details
Entrez Gene ID: | 53901 |
Gene Name: | regulator of calcineurin 2 |
Gene Aliases: |
Csp2, Dscr1l1, MCIP2, ZAKI-4 |
Location: |
Chr.17:43801281-44039516 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcan2 | NM_001286653.1 | 2 | 263 | NP_001273582.1 |
| NM_207649.1 | 2 | 396 | NP_997532.1 | |
| AB061525.1 | 2 | 263 | BAB71956.1 | |
| AK136805.1 | 2 | 204 | ||
| AK138768.1 | 2 | 308 | BAE23773.1 | |
| BB635249.1 | 2 | 308 | ||
| BY740155.1 | 2 | 403 |
More Information
Set Membership: |
|

Genomic Map