Assay Details
Target Gene Details
Entrez Gene ID: | 53901 |
Gene Name: | regulator of calcineurin 2 |
Gene Aliases: |
Csp2, Dscr1l1, MCIP2, ZAKI-4 |
Location: |
Chr.17:43801281-44039516 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcan2 | NM_001286653.1 | 3 | 379 | NP_001273582.1 |
| NM_001286654.1 | 3 | 256 | NP_001273583.1 | |
| NM_030598.2 | 2 | 386 | NP_085101.1 | |
| NM_207649.1 | 3 | 512 | NP_997532.1 | |
| AB061524.1 | 2 | 324 | BAB71955.1 | |
| AB061525.1 | 3 | 379 | BAB71956.1 | |
| AF237791.1 | 2 | 126 | AAF63487.1 | |
| AF237887.1 | 2 | 172 | AAF62538.1 | |
| AK049006.1 | 2 | 325 | BAC33511.1 | |
| AK136805.1 | 3 | 320 | ||
| AK138768.1 | 3 | 424 | BAE23773.1 | |
| AK160865.1 | 2 | 274 | BAE36057.1 | |
| BB635249.1 | 3 | 424 | ||
| BC047153.1 | 2 | 184 | AAH47153.1 | |
| BC049096.1 | 2 | 386 | AAH49096.1 | |
| BC062141.1 | 3 | 256 | AAH62141.1 | |
| BY740155.1 | 3 | 519 |
More Information
Set Membership: |
|

Genomic Map