Assay Details
Target Gene Details
Entrez Gene ID: | 53901 |
Gene Name: | regulator of calcineurin 2 |
Gene Aliases: |
Csp2, Dscr1l1, MCIP2, ZAKI-4 |
Location: |
Chr.17:43801281-44039516 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcan2 | NM_001286653.1 | 4 | 641 | NP_001273582.1 |
| NM_001286654.1 | 4 | 518 | NP_001273583.1 | |
| NM_030598.2 | 3 | 648 | NP_085101.1 | |
| NM_207649.1 | 4 | 774 | NP_997532.1 | |
| AB061524.1 | 3 | 586 | BAB71955.1 | |
| AB061525.1 | 4 | 641 | BAB71956.1 | |
| AF237791.1 | 3 | 388 | AAF63487.1 | |
| AF237887.1 | 3 | 434 | AAF62538.1 | |
| AK049006.1 | 3 | 587 | BAC33511.1 | |
| AK136805.1 | 4 | 582 | ||
| AK138768.1 | 4 | 686 | BAE23773.1 | |
| AK160865.1 | 3 | 536 | BAE36057.1 | |
| BB635249.1 | ||||
| BC047153.1 | 3 | 446 | AAH47153.1 | |
| BC049096.1 | 3 | 648 | AAH49096.1 | |
| BC062141.1 | 4 | 518 | AAH62141.1 |
More Information
Set Membership: |
|

Genomic Map