Assay Details
Target Gene Details
Entrez Gene ID: | 27632 |
Gene Name: | negative elongation factor complex member E, Rdbp |
Gene Aliases: |
D17H6S45, NELF-E, RD, Rdbp |
Location: |
Chr.17:34850391-34856372 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nelfe | NM_001045863.1 | 10 | 1185 | NP_001039328.1 |
| NM_001045864.1 | 10 | 1271 | NP_001039329.1 | |
| NM_138580.2 | 10 | 1059 | NP_613046.2 | |
| AI325150.1 | 2 | 351 | ||
| AK011663.1 | 10 | 1044 | ||
| AK012389.1 | 10 | 1076 | BAB28206.1 | |
| AK131947.1 | 10 | 1043 | BAE20893.1 | |
| AK147011.1 | 10 | 1167 | BAE27605.1 | |
| BC034867.1 | 10 | 1011 | AAH34867.1 | |
| DV662511.1 |

Genomic Map