Assay Details
Target Gene Details
Entrez Gene ID: | 14670 |
Gene Name: | guanine nucleotide binding protein-like 1 |
Gene Aliases: |
Gna-rs1, Gnal1 |
Location: |
Chr.17:35979955-35989462 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gnl1 | NM_008136.2 | 4 | 671 | NP_032162.2 |
AK078187.1 | 4 | 632 | ||
BC010298.1 | 4 | 626 | AAH10298.1 | |
BC094391.1 | 2 | 108 | AAH94391.1 | |
BC129969.1 | 2 | 203 | ||
BC129970.1 | 2 | 203 | ||
BM949933.1 | 4 | 417 | ||
X65026.1 | 3 | 314 | CAA46160.1 |
