Assay Details
Target Gene Details
Entrez Gene ID: | 18207 |
Gene Name: | nth (endonuclease III)-like 1 (E.coli) |
Gene Aliases: |
Nth1, Octs3 |
Location: |
Chr.17:24632682-24638838 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nthl1 | NM_008743.2 | 2 | 256 | NP_032769.2 |
| AB006812.1 | 2 | 202 | BAA22080.1 | |
| AK033701.1 | 2 | 1264 | ||
| BC138852.1 | 2 | 219 | ||
| BC138855.1 | 2 | 219 | ||
| BC145443.1 | 2 | 128 | ||
| CX565655.1 | 2 | 235 | ||
| Y09688.1 | 2 | 228 | CAA70866.1 |
More Information
Set Membership: |
|

Genomic Map