Assay Details
Target Gene Details
Entrez Gene ID: | 225049 |
Gene Name: | tetratricopeptide repeat domain 7 |
Gene Aliases: |
1110035E02Rik, 1700007L07Rik, Ttc7a, fsn, hea |
Location: |
Chr.17:87282082-87381770 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttc7 | NM_028639.3 | 7 | 1264 | NP_082915.1 |
| AK040578.1 | 7 | 1267 | BAC30634.1 | |
| AK040610.1 | 7 | 1235 | BAC30643.1 | |
| AK050263.1 | 7 | 1251 | BAC34153.1 | |
| AK080284.1 | 7 | 1243 | BAC37865.1 | |
| AK153704.1 | 7 | 1246 | ||
| BC042512.1 | 7 | 941 | AAH42512.2 |

Genomic Map