Assay Details
Target Gene Details
Entrez Gene ID: | 338363 |
Gene Name: | transmembrane protein 241 |
Gene Aliases: |
6030446N20Rik |
Location: |
Chr.18:11980136-12121543 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem241 | NM_001289666.1 | NP_001276595.1 | ||
| NM_001289667.1 | NP_001276596.1 | |||
| NM_178801.5 | NP_848916.2 | |||
| NR_110356.1 | ||||
| AK031517.1 | BAC27431.1 | |||
| AK038419.1 | BAC29994.1 | |||
| AK038472.1 | BAC30011.1 | |||
| AK144963.1 | BAE26156.1 | |||
| BC094382.1 | AAH94382.1 | |||
| BC137621.1 |
More Information
Set Membership: |
|

Genomic Map