Assay Details
Target Gene Details
Entrez Gene ID: | 106489 |
Gene Name: | SFT2 domain containing 1 |
Gene Aliases: |
5630401J11Rik, AA409698 |
Location: |
Chr.17:8311103-8327442 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sft2d1 | NM_134114.2 | NP_598875.2 | ||
| AK017471.1 | ||||
| AK087027.1 | ||||
| AK151338.1 | BAE30316.1 | |||
| AK171776.1 | BAE42659.1 | |||
| BC091770.1 | AAH91770.1 | |||
| BC130232.1 |
More Information
Set Membership: |
|

Genomic Map