Assay Details
Target Gene Details
Entrez Gene ID: | 20405 |
Gene Name: | SH3-domain GRB2-like 1 |
Gene Aliases: |
C77078, EEN, SH3P8, Sh3d2b |
Location: |
Chr.17:56016750-56036638 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sh3gl1 | NM_001252471.1 | NP_001239400.1 | ||
| NM_013664.3 | NP_038692.1 | |||
| AK154836.1 | BAE32864.1 | |||
| AK162715.1 | BAE37032.1 | |||
| AK167505.1 | BAE39581.1 | |||
| BC138602.1 | ||||
| BC138603.1 | ||||
| U58885.1 | AAC71775.1 |

Genomic Map