Assay Details
Target Gene Details
Entrez Gene ID: | 20420 |
Gene Name: | src homology 2 domain-containing transforming protein D |
Gene Aliases: |
AI413439 |
Location: |
Chr.17:55968392-55976625 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shd | NM_001159523.1 | NP_001152995.1 | ||
| NM_009168.2 | NP_033194.1 | |||
| AB018423.1 | BAA33805.1 | |||
| AK142491.1 | BAE25084.1 | |||
| AK164639.1 | BAE37855.1 | |||
| BC103798.1 | AAI03799.1 |

Genomic Map