Assay Details
Target Gene Details
Entrez Gene ID: | 65962 |
Gene Name: | solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 2 |
Gene Aliases: |
0610011L07Rik, 1200011K07Rik, 2010007A20Rik, E3karp, NHERF-2, Nherf2, Octs2, Sip-1, Sip1, Sryip1, Tka-1 |
Location: |
Chr.17:24639281-24650340 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc9a3r2 | NM_023055.2 | NP_075542.2 | ||
| NM_023449.3 | NP_075938.2 | |||
| AB026489.1 | BAA97568.1 | |||
| AK002557.1 | BAB22185.1 | |||
| AK004710.1 | ||||
| AK039491.1 | BAC30366.1 | |||
| AK075800.1 | ||||
| AK075813.1 | BAC35980.1 | |||
| AK084801.1 | BAC39282.1 | |||
| AK169007.1 | ||||
| AK170057.1 | BAE41537.1 | |||
| AK179335.1 | ||||
| BC065778.1 | AAH65778.1 | |||
| BC070458.1 | AAH70458.1 |
More Information
Set Membership: |
|

Genomic Map