Assay Details
Target Gene Details
Entrez Gene ID: | 22436 |
Gene Name: | xanthine dehydrogenase |
Gene Aliases: |
XO, Xor, Xox-1, Xox1 |
Location: |
Chr.17:73883895-73950196 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xdh | NM_011723.3 | 14 | 1458 | NP_035853.2 |
| AK034256.1 | 14 | 1408 | ||
| AK090058.1 | 5 | 599 | ||
| AK143412.1 | 14 | 1406 | ||
| AK144700.1 | 14 | 1410 | BAE26022.1 | |
| AK160010.1 | 11 | 1157 | ||
| AK161529.1 | 14 | 1409 | ||
| BC141183.1 | 14 | 1433 | ||
| X62932.1 | 14 | 1392 | CAA44705.1 |
More Information
Set Membership: |
|

Genomic Map