Assay Details
Target Gene Details
Entrez Gene ID: | 56468 |
Gene Name: | suppressor of cytokine signaling 5 |
Gene Aliases: |
1810018L08Rik, Cish5, mKIAA0671 |
Location: |
Chr.17:87107407-87137647 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Socs5 | NM_019654.2 | NP_062628.2 | ||
| AK048342.1 | ||||
| AK135099.1 | BAE22422.1 | |||
| BC053015.1 | AAH53015.1 |
More Information
Set Membership: |
|

Genomic Map