Assay Details
Target Gene Details
Entrez Gene ID: | 225289 |
Gene Name: | expressed sequence AW554918 |
Gene Aliases: |
4921538P22, AI461742, Kiaa1328, mKIAA1328 |
Location: |
Chr.18:25169004-25467321 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AW554918 | NM_001033532.3 | NP_001028704.2 | ||
| AK167257.1 | BAE39373.1 |

Genomic Map