Assay Details
Target Gene Details
Entrez Gene ID: | 225372 |
Gene Name: | amyloid beta (A4) precursor protein-binding, family B, member 3 |
Gene Aliases: |
Fe65l2, Rirl2, TR2S |
Location: |
Chr.18:36671154-36679366 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apbb3 | NM_146085.1 | NP_666197.1 | ||
| AK141878.1 | BAE24865.1 | |||
| AK171634.1 | ||||
| AK177880.1 | ||||
| AK210931.1 | ||||
| BC024809.1 | AAH24809.1 |

Genomic Map