Assay Details
Target Gene Details
Entrez Gene ID: | 12475 |
Gene Name: | CD14 antigen |
Gene Aliases: |
- |
Location: |
Chr.18:36725064-36726815 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd14 | NM_009841.4 | 1 | 237 | NP_033971.1 |
| AK149744.1 | 1 | 164 | BAE29057.1 | |
| AK152427.1 | ||||
| AK153229.1 | 1 | 27 | BAE31822.1 | |
| AK156262.1 | BAE33646.1 | |||
| BC057889.1 | 1 | 76 | AAH57889.1 | |
| X13333.1 | CAA31710.1 |
More Information
Set Membership: |
|

Genomic Map