Assay Details
Target Gene Details
Entrez Gene ID: | 93687 |
Gene Name: | casein kinase 1, alpha 1 |
Gene Aliases: |
2610208K14Rik, 4632404G05Rik, 5430427P18Rik, CK1a, Csnk1a |
Location: |
Chr.18:61555165-61590315 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csnk1a1 | NM_146087.2 | NP_666199.1 | ||
| AK019176.1 | 2 | 670 | ||
| AK076070.1 | BAC36161.1 | |||
| AK076264.1 | ||||
| AK078398.1 | BAC37255.1 | |||
| AK141421.1 | ||||
| AK146873.1 | BAE27496.1 | |||
| AK147165.1 | BAE27730.1 | |||
| AK152387.1 | BAE31175.1 | |||
| AK152793.1 | BAE31499.1 | |||
| AK166318.1 | BAE38701.1 | |||
| AK166429.1 | ||||
| AK167550.1 | ||||
| AK210280.1 | ||||
| BC019740.1 | AAH19740.1 | |||
| BC025439.1 | AAH25439.1 | |||
| BC048081.1 | AAH48081.1 |
More Information
Set Membership: |
|

Genomic Map