Assay Details
Target Gene Details
Entrez Gene ID: | 81877 |
Gene Name: | tenascin XB |
Gene Aliases: |
Tn-mhc, Tnx |
Location: |
Chr.17:34659788-34795460 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnxb | NM_031176.2 | 25 | 8239 | NP_112453.2 |
| AB235155.1 | 25 | 8239 | BAE44473.1 | |
| AK136535.1 | 13 | 3335 | ||
| AK171696.1 | 12 | 2974 | BAE42616.1 | |
| X73959.1 | 7 | 1441 | CAA52147.1 |
More Information
Set Membership: |
|

Genomic Map